ENA018110474 Enamine

[(4-fluorophenyl)(phenyl)methyl](methyl)amine

  • MDL number MFCD06660684
  • Cas Number : 885459-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07498
Availability details for
Quantity
  • Availability
  • Add to Cart