ENA018110486 Enamine

3-(4-methylphenyl)-2-sulfanylidene-1,3-thiazolidin-4-one

  • MDL number MFCD00268237
  • Cas Number : 3919-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07510
Availability details for
Quantity
  • Availability
  • Add to Cart