ENA018110491 Enamine

3-(3-methylbutyl)-2-sulfanyl-5-(thiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD04632088
  • Cas Number : 731827-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07515

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07515
Availability details for
Quantity
  • Availability
  • Add to Cart