ENA018110498 Enamine

4-(4-chlorophenyl)-5-(3-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD04632074
  • Cas Number : 731827-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07522
Availability details for
Quantity
  • Availability
  • Add to Cart