ENA018110505 Enamine

7-chloro-3-(2-methylpropyl)-2-sulfanyl-3,4-dihydroquinazolin-4-one

  • MDL number MFCD04632090
  • Cas Number : 731827-21-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221-222
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07529

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07529
Availability details for
Quantity
  • Availability
  • Add to Cart