ENA018110507 Enamine

11-cyclopentyl-10-sulfanyl-7-thia-9,11-diazatricyclo[6.4.0.0,2,6]dodeca-1(8),2(6),9-trien-12-one

  • MDL number MFCD04632061
  • Cas Number : 731827-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07531

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07531
Availability details for
Quantity
  • Availability
  • Add to Cart