ENA018110514 Enamine

3-(2-methylphenyl)-5-phenyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD04632134
  • Cas Number : 885459-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07538

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07538
Availability details for
Quantity
  • Availability
  • Add to Cart