ENA018110517 Enamine

3-isobutyl-2-mercapto-5-phenylthieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD04632077
  • Cas Number : 731827-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07541

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07541
Availability details for
Quantity
  • Availability
  • Add to Cart