ENA018110527 Enamine

3-{[(1-adamantylamino)carbonyl]amino}propanoic acid

  • MDL number MFCD01791133
  • Cas Number : 33205-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07551

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07551
Availability details for
Quantity
  • Availability
  • Add to Cart