ENA018110532 Enamine

bis(4-phenylphenyl)ethane-1,2-dione

  • MDL number MFCD01142538
  • Cas Number : 4746-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07556

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07556
Availability details for
Quantity
  • Availability
  • Add to Cart