ENA018110533 Enamine

1,2-bis({[1,1'-biphenyl]-4-yl})-2-hydroxyethan-1-one

  • MDL number MFCD01454300
  • Cas Number : 5623-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07557
Availability details for
Quantity
  • Availability
  • Add to Cart