ENA018110547 Enamine

4-(2-methoxyethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06654962
  • Cas Number : 76457-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07571

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07571
Availability details for
Quantity
  • Availability
  • Add to Cart