ENA018110550 Enamine

4-(3-methoxypropyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06654964
  • Cas Number : 75028-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07574
Availability details for
Quantity
  • Availability
  • Add to Cart