ENA018110554 Enamine

4-[(4-methylphenyl)methoxy]benzaldehyde

  • MDL number MFCD00617729
  • Cas Number : 66742-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07578
Availability details for
Quantity
  • Availability
  • Add to Cart