ENA018110567 Enamine

6-amino-5-(2-chloropropanoyl)-3-methyl-1-propylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06660489
  • Cas Number : 851169-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07592

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07592
Availability details for
Quantity
  • Availability
  • Add to Cart