ENA018110572 Enamine

2-{[5-(cyclohexylamino)-1,3,4-thiadiazol-2-yl]sulfanyl}acetic acid

  • MDL number MFCD04632076
  • Cas Number : 68161-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07598

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07598
Availability details for
Quantity
  • Availability
  • Add to Cart