ENA018110580 Enamine

7-hexyl-5-mercapto-3-methyl-1-propylpyrimido[4,5-d]pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06654969
  • Cas Number : 851169-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172...174
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07606

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07606
Availability details for
Quantity
  • Availability
  • Add to Cart