ENA018110582 Enamine

3,7-dimethyl-1-(2-methylpropyl)-5-sulfanyl-1H,2H,3H,4H-[1,3]diazino[4,5-d]pyrimidine-2,4-dione

  • MDL number MFCD06660490
  • Cas Number : 794559-23-2
  • Molecular Weight : 280.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07608
Availability details for
Quantity
  • Availability
  • Add to Cart