ENA018110585 Enamine

N-[4-(2-chlorophenyl)-1,3-thiazol-2-yl]-2-cyanoacetamide

  • MDL number MFCD06654972
  • Cas Number : 851169-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07611
Availability details for
Quantity
  • Availability
  • Add to Cart