ENA018110591 Enamine

5-ethyl-4-methylthiophene-2-carboxylic acid

  • MDL number MFCD06654975
  • Cas Number : 40808-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98-99
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07617
Availability details for
Quantity
  • Availability
  • Add to Cart