ENA018110608 Enamine

7-bromo-1,2,3,4-tetrahydroacridine-9-carboxylic acid

  • MDL number MFCD00681032
  • Cas Number : 37509-14-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 291...293
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07634

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07634
Availability details for
Quantity
  • Availability
  • Add to Cart