ENA018110614 Enamine

2-hydrazino-7-methoxy-4-methylquinoline

  • MDL number MFCD00711758
  • Cas Number : 97892-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250...252
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07640

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07640
Availability details for
Quantity
  • Availability
  • Add to Cart