ENA018110632 Enamine

2-(2-bromo-4-formyl-6-methoxyphenoxy)-N,N-dimethylacetamide

  • MDL number MFCD04633430
  • Cas Number : 732291-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07659
Availability details for
Quantity
  • Availability
  • Add to Cart