ENA024293714 Enamine

4-sec-butylbenzenesulfonyl chloride

  • MDL number MFCD00024886
  • Cas Number : 56609-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07674

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-07674
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart