ENA024293732 Enamine

2-chloro-N-(2-chloro-5-methanesulfonylphenyl)acetamide

  • MDL number MFCD06364986
  • Cas Number : 793727-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07692

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07692
Availability details for
Quantity
  • Availability
  • Add to Cart