ENA024293753 Enamine

2-methyl-5-(thiophen-2-yl)thieno[2,3-d]pyrimidine-4-thiol

  • MDL number MFCD04633474
  • Cas Number : 732292-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07713
Availability details for
Quantity
  • Availability
  • Add to Cart