ENA024293755 Enamine

2-sulfanyl-5-(thiophen-2-yl)-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03815809
  • Cas Number : 372494-00-3
  • Molecular Weight : 266.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07715
Availability details for
Quantity
  • Availability
  • Add to Cart