ENA024293759 Enamine

2-(ethylamino)-N-(4-methylphenyl)propanamide

  • MDL number MFCD04629595
  • Cas Number : 1008058-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07719
Availability details for
Quantity
  • Availability
  • Add to Cart