ENA024293768 Enamine

1-[(2-chloroethyl)sulfanyl]-4-methylbenzene

  • MDL number MFCD00093775
  • Cas Number : 20761-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07728

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07728
Availability details for
Quantity
  • Availability
  • Add to Cart