ENA024293790 Enamine

2-(4,4-diethyl-2,5-dioxoimidazolidin-1-yl)acetic acid

  • MDL number MFCD06654997
  • Cas Number : 714-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-171
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07750
Availability details for
Quantity
  • Availability
  • Add to Cart