ENA024293801 Enamine

6-fluoro-3,4-dihydro-2H-1lambda6-benzothiopyran-1,1,4-trione

  • MDL number MFCD00728617
  • Cas Number : 21243-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-177
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07761
Availability details for
Quantity
  • Availability
  • Add to Cart