ENA024293803 Enamine

2-cyano-N-[4-(3,4-difluorophenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD06655000
  • Cas Number : 851169-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...180
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07763

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07763
Availability details for
Quantity
  • Availability
  • Add to Cart