ENA024293805 Enamine

N-(4-tert-butyl-1,3-thiazol-2-yl)-2-cyanoacetamide

  • MDL number MFCD06655001
  • Cas Number : 851169-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07765

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07765
Availability details for
Quantity
  • Availability
  • Add to Cart