ENA024293811 Enamine

3-(morpholine-4-carbonyl)naphthalen-2-ol

  • MDL number MFCD01017081
  • Cas Number : 3692-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07771
Availability details for
Quantity
  • Availability
  • Add to Cart