ENA024293821 Enamine

4-amino-5-(2-bromophenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD02046316
  • Cas Number : 61055-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07781

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07781
Availability details for
Quantity
  • Availability
  • Add to Cart