ENA024293823 Enamine

3-chloro-5-ethoxy-4-hydroxybenzoic acid

  • MDL number MFCD04635820
  • Cas Number : 733030-51-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07783

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07783
Availability details for
Quantity
  • Availability
  • Add to Cart