ENA024293830 Enamine

1-(3,5-dimethylphenyl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD04633098
  • Cas Number : 890010-89-6
  • Molecular Weight : 201.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-79
Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-07790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07790
Availability details for
Quantity
  • Availability
  • Add to Cart