ENA024293841 Enamine

1-ethyl-5-(morpholine-4-sulfonyl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD04635810
  • Cas Number : 733030-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07801
Availability details for
Quantity
  • Availability
  • Add to Cart