ENA024293843 Enamine

3-[5-(dimethylsulfamoyl)-1-methyl-1H-1,3-benzodiazol-2-yl]propanoic acid

  • MDL number MFCD04635811
  • Cas Number : 733030-45-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 207-208
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07803
Availability details for
Quantity
  • Availability
  • Add to Cart