ENA024293846 Enamine

1-[4-(2-methylpropyl)phenyl]ethan-1-amine

  • MDL number MFCD05215266
  • Cas Number : 74305-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07806
Availability details for
Quantity
  • Availability
  • Add to Cart