ENA024293847 Enamine

5-chloro-2-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]aniline

  • MDL number MFCD04635839
  • Cas Number : 733030-65-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07807
Availability details for
Quantity
  • Availability
  • Add to Cart