ENA024293852 Enamine

[({4-[(2-furylmethyl)amino]-3-nitrophenyl}sulfonyl)amino]acetic acid

  • MDL number MFCD04635859
  • Cas Number : 733030-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07812

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07812
Availability details for
Quantity
  • Availability
  • Add to Cart