ENA024293857 Enamine

[2-(diethylamino)ethyl]thiourea

  • MDL number MFCD04635869
  • Cas Number : 66892-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-07817

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07817
Availability details for
Quantity
  • Availability
  • Add to Cart