ENA024293866 Enamine

ethyl 2-amino-5-methyl-4-(2-methylbutyl)thiophene-3-carboxylate

  • MDL number MFCD04635882
  • Cas Number : 733030-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07826
Availability details for
Quantity
  • Availability
  • Add to Cart