ENA024293870 Enamine

2-(morpholin-4-ylmethyl)-3-(2-oxopropyl)quinazolin-4(3H)-one

  • MDL number MFCD04619533
  • Cas Number : 727718-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07830

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07830
Availability details for
Quantity
  • Availability
  • Add to Cart