ENA024293885 Enamine

4-bromo-1-(3,5-dimethylphenyl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD04633150
  • Cas Number : 956197-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07845

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07845
Availability details for
Quantity
  • Availability
  • Add to Cart