ENA024293899 Enamine

5-bromo-2-(difluoromethoxy)benzaldehyde

  • MDL number MFCD02709888
  • Cas Number : 329269-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07859
Availability details for
Quantity
  • Availability
  • Add to Cart