ENA024293902 Enamine

{[4-ethyl-5-(2-furyl)-4H-1,2,4-triazol-3-yl]thio}acetic acid

  • MDL number MFCD02596624
  • Cas Number : 537016-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07862

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07862
Availability details for
Quantity
  • Availability
  • Add to Cart