ENA024293910 Enamine

1-butylimidazolidine-2,4,5-trione

  • MDL number MFCD04635925
  • Cas Number : 40408-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-07870

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-07870
Availability details for
Quantity
  • Availability
  • Add to Cart