ENA024293932 Enamine

5-chloro-3-methyl-1-(4-methylbenzyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD06335051
  • Cas Number : 882140-34-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-07892

Safety Information

msds https://www.enaminestore.com/catalog/EN300-07892
Availability details for
Quantity
  • Availability
  • Add to Cart